Learn More
Thermo Scientific Kit, PCR Cloning; Thermo Scientific; InsTAclone; TA system; Fast cloning; Direct one-step procedure; High yield of target DNA; Compatible w/Taq, Tth, Tfl, other non-proofreading DNA pol, long PCR enzyme mix; For 10 reactions
Brand:  Thermo Scientific K1213
Kit, PCR Cloning; Thermo Scientific; InsTAclone; TA system; Fast cloning; Direct one-step procedure; High yield of target DNA; Compatible w/Taq, Tth, Tfl, other non-proofreading DNA pol, long PCR enzyme mix; For 10 reactions
Code : A0
Additional Details : Weight : 0.00900kg
Product Code. 10181080