Learn More
Thermo Scientific™ Biotin DecaLabel DNA Labeling Kit
Efficiently synthesize biotin-labeled DNA probes with the Thermo Scientific™ Biotin DecaLabel DNA Labeling Kit.
Brand: Thermo Scientific™ K0652
Code : 88
Additional Details : Weight : 1.58760kg
Description
Biotin DecaLabel DNA Labeling Kit is an advanced system for the efficient synthesis of biotin-labeled DNA probes, based on an improved random-primed labeling method developed by Feinberg and Vogelstein. The primary improvement over traditional random-primed method involves the use of random decamers instead of hexamers, ensuring more efficient annealing with DNA at 37°C. Klenow Fragment, exo-, included in the kit, is genetically engineered enzyme with no detectable exonuclease activity. The enzyme does not degrade the labeled probe during reaction, which results in a high labeling yield even with low amounts of template. You can uniformly label any length DNA fragments.
Biotin-labeled DNA is detected with the Biotin Chromogenic Detection Kit or conventional biotin-avidin or biotin-streptavidin detection systems.
The primary improvement over traditional random-primed method involves the use of random decamers instead of hexamers to ensure more efficient annealing with DNA at 37°C. Klenow Fragment, exo- is also included in the kit; this genetically engineered enzyme has no detectable exonuclease activity. Therefore, the enzyme does not degrade the labeled probe during reaction, which results in a high labeling yield even with low amounts of template. As a result, DNA fragments of any length can be uniformly labeled. Biotin-labeled DNA is detected with the Biotin Chromogenic Detection Kit or with conventional biotin-avidin or biotin-streptavidin detection systems.- Non-radioactive labeling of DNA
- Efficient priming of labeling reactions with random decamers
- High yields with Klenow Fragment, exo-: no degradation of a labeled probe during reaction
Recommended for:
Generation of biotin-labeled DNA probes for a variety of non-radioactive hybridization experiments, including Southern and Northern blots, colony/plaque hybridizations, dot/slot blots and in situ hybridizations
Specifications
Biotin-based | |
DNA | |
Yes | |
30 | |
DNA Labeling Kit |
Kit | |
Genomic DNA, PCR Products, DNA (General) | |
Direct Labeling | |
DecaLabel | |
30 Reactions |