Learn More
Fenretinide, Tocris Bioscience™
Synthetic retinoid; antiproliferative and antioxidant
Brand:  Tocris (Bio-Techne) 139650
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13459303
  • £507.02 / 50mg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
Synthetic retinoid agonist. Antiproliferative, antioxidant and anticancer agent with a long half-life in vivo. Apoptotic effects appear to be mediated by a mechanism distinct from that of «classical« retinoids.
Specifications
Chemical Identifiers
65646-68-6
391.555
fenretinide, n-4-hydroxyphenyl retinamide, 4-hpr, 4-hydroxyphenylretinamide, 4-hydroxyphenyl retinamide, retinoic acid p-hydroxyanilide, all-trans-4'-hydroxyretinanilide, fenretinida, fenretinidum, fenretinidum latin
CHEBI:42588
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)NC2=CC=C(C=C2)O)C)C
C26H33NO2
AKJHMTWEGVYYSE-FXILSDISSA-N
5288209
(2E,4E,6E,8E)-N-(4-hydroxyphenyl)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenamide
Specifications
Fenretinide
65646-68-6
fenretinide, n-4-hydroxyphenyl retinamide, 4-hpr, 4-hydroxyphenylretinamide, 4-hydroxyphenyl retinamide, retinoic acid p-hydroxyanilide, all-trans-4'-hydroxyretinanilide, fenretinida, fenretinidum, fenretinidum latin
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)NC2=CC=C(C=C2)O)C)C
391.555
CHEBI:42588
>98%
50mg
C26H33NO2
AKJHMTWEGVYYSE-FXILSDISSA-N
(2E,4E,6E,8E)-N-(4-hydroxyphenyl)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenamide
5288209
391.55
Videos