missing translation for 'onlineSavingsMsg'
Learn More

Fenoldopam hydrochloride, Tocris Bioscience™

Selective D1-like partial agonist

Brand:  Tocris (Bio-Techne) 165910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13439873

  • £165.00 / 10mg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective D1-like dopamine receptor partial agonist (EC50 = 57 nM). Vasodilator in vivo and does not readily cross the blood-brain barrier. Also α2-adrenoceptor antagonist in vitro (Ki = 15 - 25 nM).
Specifications

Chemical Identifiers

181217-39-0
342.216
fenoldopam hydrochloride, 6-chloro-2,3,4,5-tetrahydro-1-4-hydroxyphenyl-1h-3-benzazepine-7,8-diol hydrochloride, skf 82526 hydrochloride, inverted exclamation marka-fenoldopam hydrochloride, inverted exclamation marka-skf 82526 hydrochloride, 6-chloro-1-4-hydroxyphenyl-2,3,4,5-tetrahydro-1h-3-benzazepine-7,8-diol hydrochloride, 1h-3-benzazepine-7,8-diol, 6-chloro-2,3,4,5-tetrahydro-1-4-hydroxyphenyl-, hydrochloride 1:1
9-chloro-5-(4-hydroxyphenyl)-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol;hydrochloride
C16H17Cl2NO3
NLMPGIXLXSPNFS-UHFFFAOYSA-N
21642479
C1CNCC(C2=CC(=C(C(=C21)Cl)O)O)C3=CC=C(C=C3)O.Cl

Specifications

Fenoldopam hydrochloride
181217-39-0
fenoldopam hydrochloride, 6-chloro-2,3,4,5-tetrahydro-1-4-hydroxyphenyl-1h-3-benzazepine-7,8-diol hydrochloride, skf 82526 hydrochloride, inverted exclamation marka-fenoldopam hydrochloride, inverted exclamation marka-skf 82526 hydrochloride, 6-chloro-1-4-hydroxyphenyl-2,3,4,5-tetrahydro-1h-3-benzazepine-7,8-diol hydrochloride, 1h-3-benzazepine-7,8-diol, 6-chloro-2,3,4,5-tetrahydro-1-4-hydroxyphenyl-, hydrochloride 1:1
C1CNCC(C2=CC(=C(C(=C21)Cl)O)O)C3=CC=C(C=C3)O.Cl
342.216
342.22
10mg
C16H17Cl2NO3
NLMPGIXLXSPNFS-UHFFFAOYSA-N
9-chloro-5-(4-hydroxyphenyl)-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol;hydrochloride
21642479
>98%
Videos