missing translation for 'onlineSavingsMsg'
Learn More

Ronnel, Fenchlorphos, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  Paragon Scientific S2000

Code : 50

Additional Details : Weight : 0.80000kg

Product Code. 11939768

  • £56.25 / Each


Estimated Shipment
01-03-2018
Add to basket

Disclaimers:

Product intended for laboratory use only


Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

AnalyteCAS No.Concentration
Fenchlorphos299-84-31000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Ronnel (Fenclorphos)
99.9
2mL amber prescored glass ampule
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
1mL
67-56-1
99.9
Fenchlorphos
Methanol
GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Videos