Learn More
Fehlings No.1 Indicator Solution, Reagecon
High-quality indicator solution produced from the highest quality raw materials and manufactured and tested under rigorous conditions.
Brand: Reagecon FS0101
Code : NEW
Additional Details : CAS Number : 7758-98-7 Weight : 1.18000kg Transport : UN number : 3082 Chem class : 9 Pack group : III
Description
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- For manual titrations; indicator titrant or analyte change color and this color change are by far the most important method of end point detection.
- This high-quality indicator solution facilitates end point detection, in a clear and unambiguous manner and is an imperative for accurate manual titrimetry.
Specifications
Accuracy ± 0.1% | |
1 L | |
Liquid |
HDPE Bottle GL45 | |
Titration & Analyte Color Change |
Safety and Handling
- FEHLINGS SOLUTION 1
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : copper sulphate