missing translation for 'onlineSavingsMsg'
Learn More

Febuxostat, Tocris Bioscience™

Xanthine oxidase inhibitor

Brand:  RD Systems (Bio-Techne) 4427/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13444734

  • £135.00 / 10mg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Non-purine inhibitor of xanthine oxidase (Ki = 1.2 nM). Binds to a channel leading to the enzyme active site; obstructs substrate binding.
Specifications

Chemical Identifiers

144060-53-7
316.375
febuxostat, adenuric, uloric, feburic, tmx 67, 2-3-cyano-4-isobutoxyphenyl-4-methylthiazole-5-carboxylic acid, tmx-67, febuxostat usan, unii-101v0r1n2e, uloric tn
CHEBI:45943
CC1=C(SC(=N1)C2=CC(=C(C=C2)OCC(C)C)C#N)C(=O)O
C16H16N2O3S
BQSJTQLCZDPROO-UHFFFAOYSA-N
134018
2-[3-cyano-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylic acid

Specifications

Febuxostat
144060-53-7
febuxostat, adenuric, uloric, feburic, tmx 67, 2-3-cyano-4-isobutoxyphenyl-4-methylthiazole-5-carboxylic acid, tmx-67, febuxostat usan, unii-101v0r1n2e, uloric tn
CC1=C(SC(=N1)C2=CC(=C(C=C2)OCC(C)C)C#N)C(=O)O
316.375
CHEBI:45943
>99%
10mg
C16H16N2O3S
BQSJTQLCZDPROO-UHFFFAOYSA-N
2-[3-cyano-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylic acid
134018
316.37
Videos