Learn More
Alfa Aesar™ FCS Blocking Buffer, in PBS
For blocking excess solid surface in ELISA assays
Brand:  Alfa Aesar™ J63160.AP
Code : NEW
Additional Details : Weight : 0.00050kg
 View more versions of this product
Product Code. 15485419
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Specifications
Specifications
For blocking excess solid surface in ELISA assays
Liquid
Blocking Buffer
500mL