missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FCGR4 (CD16-2) Recombinant Mouse Protein, His Tag

Expressed from DNA sequence encoding extracellular domain (Met 1-Gln 203) of mouse FCGR4 fused to polyhistidine tag at C-terminus

Brand:  Thermo Scientific™ 50036M08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13433129

  • £57.00 / 5µg


Estimated Shipment
12-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
246256
≥90% by SDS-PAGE
Mouse
<1 EU/μg
Fcgr4
CD Proteins
FCGR4 (CD16-2) Recombinant Mouse Protein
Lyophilized
22.4kd
5μg
Truncated, Cell Surface Protein, Recombinant
Fc gamma receptor receptor IV
Assay Standard
Mammalian
His Tag
Videos