missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FCGR4 (CD16-2) Recombinant Mouse Protein, His Tag

Expressed from DNA sequence encoding extracellular domain (Met 1-Gln 203) of mouse FCGR4 fused to polyhistidine tag at C-terminus

Brand:  Thermo Scientific™ 50036M08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13443129

  • £214.00 / 50µg

Estimated Shipment
21-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
Specifications

Specifications

Mammalian
Assay Standard
Lyophilized
22.4kd
50μg
Truncated, Cell Surface Protein, Recombinant
Fc gamma receptor receptor IV
FCGR4 (CD16-2) Recombinant Mouse Protein
CD Proteins
Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
246256
≥90% by SDS-PAGE
Mouse
<1 EU/μg
Fcgr4
His Tag
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.