Learn More
Thermo Scientific™ FCGR3A (CD16a) Recombinant Human Protein, His.AVI Tag
Expressed from DNA sequence encoding extracellular domain (Met 1-Gln 208) of human FCGR3A containing 176 Phe/Val mutation fused to a polyhistidine tag
Brand: Thermo Scientific™ 10389H27H25
Code : NEW
Additional Details : Weight : 0.00001kg
Description
- Measured using Octet RED System affinity constant (Kd) of FCGR3A bound to human IgG1 was 80nM
- Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
Specifications
Studying protein-protein interactions, and cell biology research applications | |
2214 | |
≥90% by SDS-PAGE | |
Human | |
<1 EU/μg | |
FCGR3A (CD16a) | |
CD Proteins, Growth Factors and Receptors | |
FCGR3A (CD16a) Recombinant Human Protein |
Lyophilized | |
23.3kd | |
5 x 5μg | |
Truncated, Recombinant, Cell Surface Protein | |
Fc gamma receptor III-a | |
Assay Standard | |
Mammalian | |
AVI Tag, His Tag |
For Research Use Only. Not for use in diagnostic procedures.