missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ FCGR3A (CD16a) Recombinant Human Protein, His.AVI Tag

Expressed from DNA sequence encoding extracellular domain (Met 1-Gln 208) of human FCGR3A containing 176 Phe/Val mutation fused to a polyhistidine tag

Brand:  Sino Biological™ 10389H27H5

Code : NEW

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 13472999

  • £57.00 / 5µg


Estimated Shipment
16-05-2018
Add to basket

Description

Description

  • Measured using Octet RED System affinity constant (Kd) of FCGR3A bound to human IgG1 was 80nM
  • Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
Specifications

Specifications

Studying protein-protein interactions, and cell biology research applications
2214
≥90% by SDS-PAGE
Human
<1 EU/μg
FCGR3A (CD16a)
CD Proteins, Growth Factors and Receptors
FCGR3A (CD16a) Recombinant Human Protein
Lyophilized
23.3kd
5μg
Truncated, Recombinant, Cell Surface Protein
Fc gamma receptor III-a
Assay Standard
Mammalian
AVI Tag, His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.