missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ FCGR3A (CD16a), 158 Phe/Val Recombinant Human Protein, His.AVI Tag, Biotinylated

Expressed from DNA sequence encoding extracellular domain (Met 1-Gln 208) of human FCGR3A w/176 Phe/Val mutation

Brand:  Sino Biological™ 10389H27HB5

Code : NEW

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 13413009

  • £55.44 / 5µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

  • Used to polyhistidine and AVI tags at C-terminus
  • Expressed protein was biotinylated in vivo by Biotin-Protein ligase (BirA enzyme) which co-expressed
  • Measured by its ability to bind recombinant human IgG1 in functional ELISA
Specifications

Specifications

Studying protein-protein interactions and cell biology research applications
2214
≥85% by SDS-PAGE
Human
<1 EU/μg
FCGR3A
CD Proteins, Growth Factors and Receptors
FCGR3A (CD16a), 158 Phe/Val Recombinant Human Protein
Lyophilized
25.6kd
5μg
Truncated, Recombinant, Cell Surface Protein
Fc gamma receptor III-a
Assay Standard
Mammalian
AVI Tag, His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.