Learn More
Sino Biological™ FCGR2B (CD32b) Recombinant Human Protein, His Tag
Expressed from DNA sequence encoding extracellular domain (Ala 46-Pro 217) of human FCGR2B w/N-terminal signal peptide, C-terminal polyhistidine tag
Brand: Sino Biological™ 10259H08H25
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Measured by its binding ability in functional ELISA
- Immobilized recombinant human FCGR2B at 10μg/mL binds human IgG2 w/linear range of 0.16-6.4μg/mL
- Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
Specifications
Analysis of protein structure, studying protein-protein interactions and cell biology research | |
2213 | |
≥90% by SDS-PAGE | |
Human | |
Truncated, Recombinant, Cell Surface Protein | |
Fc gamma receptor II-b | |
Assay Standard | |
Mammalian | |
His Tag |
Lyophilized | |
20.8kd | |
5 x 5μg | |
−20° to −70°C | |
<1 EU/μg | |
FCGR2B | |
CD Proteins, Growth Factors and Receptors | |
FCGR2B (CD32b) Recombinant Human Protein |
For Research Use Only. Not for use in diagnostic procedures.