missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ FCGR2B (CD32b) Recombinant Human Protein, His Tag

Expressed from DNA sequence encoding extracellular domain (Ala 46-Pro 217) of human FCGR2B w/N-terminal signal peptide, C-terminal polyhistidine tag

Brand:  Sino Biological™ 10259H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13422959

  • £56.30 / 5µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

  • Measured by its binding ability in functional ELISA
  • Immobilized recombinant human FCGR2B at 10μg/mL binds human IgG2 w/linear range of 0.16-6.4μg/mL
  • Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
Specifications

Specifications

Analysis of protein structure, studying protein-protein interactions and cell biology research
2213
≥90% by SDS-PAGE
Human
Truncated, Recombinant, Cell Surface Protein
Fc gamma receptor II-b
Assay Standard
Mammalian
His Tag
Lyophilized
20.8kd
5μg
−20° to −70°C
<1 EU/μg
FCGR2B
CD Proteins, Growth Factors and Receptors
FCGR2B (CD32b) Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.