missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ FCGR2A (CD32a) Recombinant Human Protein

Expressed from DNA sequence encoding extracellular domain (Met 1-Gly 216) of human FCGR2A precursor fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 10374H08B50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13455846

  • £213.41 / 50µg


Estimated Shipment
29-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Lyophilized in 20mM Tris buffer, 50mM NaCl, pH 7.4
  • Contains 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
Specifications

Specifications

Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
2212
≥85% by SDS-PAGE
Human
<1 EU/μg
FCGR2A
CD Proteins
FCGR2A (CD32a) Recombinant Human Protein
Lyophilized
22kd
50μg
Truncated, Recombinant, Cell Surface Protein
Fc gamma receptor II-a
Assay Standard
Insect
His Tag
Videos