missing translation for 'onlineSavingsMsg'
Learn More

FCGR2A (CD32a) Recombinant Human Protein, Invitrogen™ Sino Biological™

Expressed from DNA sequence encoding extracellular domain (Met 1-Gly 216) of human FCGR2A precursor fused to polyhistidine tag at C-terminus

Brand:  Invitrogen 10374H08B250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13435846

  • £765.00 / 250µg

Estimated Shipment
21-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Lyophilized in 20mM Tris buffer, 50mM NaCl, pH 7.4
  • Contains 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
Specifications

Specifications

Insect
Assay Standard
Lyophilized
22kd
5 x 50μg
Truncated, Recombinant, Cell Surface Protein
Fc gamma receptor II-a
FCGR2A (CD32a) Recombinant Human Protein
CD Proteins
Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
2212
≥85% by SDS-PAGE
Human
<1 EU/μg
FCGR2A
His Tag
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.