missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FCGR2A (CD32a) Recombinant Human Protein

Expressed from DNA sequence encoding extracellular domain (Met 1-Gly 216) of human FCGR2A precursor fused to polyhistidine tag at C-terminus

Brand:  Thermo Scientific™ 10374H08B25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13425846

  • £141.58 / 25µg


Estimated Shipment
12-06-2018
Add to basket

Description

Description

  • Lyophilized in 20mM Tris buffer, 50mM NaCl, pH 7.4
  • Contains 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
Specifications

Specifications

Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
2212
≥85% by SDS-PAGE
Human
<1 EU/μg
FCGR2A
CD Proteins
FCGR2A (CD32a) Recombinant Human Protein
Lyophilized
22kd
5 x 50μg
Truncated, Recombinant, Cell Surface Protein
Fc gamma receptor II-a
Assay Standard
Insect
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.