Learn More
FCCP, Tocris Bioscience™
Oxidative phosphorylation uncoupler
Brand:  Tocris (Bio-Techne) 045310
Code : Z1
Additional Details : Weight : 0.01100kg
Quantity 10mg
 View more versions of this product
Product Code. 13457663
  • £135.24 / 10mg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
A very potent uncoupler of oxidative phosphorylation in mitochondria.
Specifications
Chemical Identifiers
370-86-5
254.172
fccp, carbonyl cyanide p-trifluoromethoxyphenylhydrazone, carbonyl cyanide 4-trifluoromethoxy phenylhydrazone, 2-2-4-trifluoromethoxy phenyl hydrazono malononitrile, propanedinitrile, 4-trifluoromethoxy phenyl hydrazono, 1-cyano-n-4-trifluoromethoxy phenyl methanecarbohydrazonoyl cyanide, carbonyl cyanide 4-trifluoromethoxyphenylhydrazone, carbonyl cyanide, 4-trifluoromethoxyphenylhydrazone, 4-trifluoromethoxy phenyl hydrazono malononitrile, mesoxalonitrile, p-trifluoromethoxy phenyl hydrazone
CHEBI:75458
C1=CC(=CC=C1NN=C(C#N)C#N)OC(F)(F)F
C10H5F3N4O
BMZRVOVNUMQTIN-UHFFFAOYSA-N
3330
2-[[4-(trifluoromethoxy)phenyl]hydrazinylidene]propanedinitrile
Specifications
10mg
370-86-5
fccp, carbonyl cyanide p-trifluoromethoxyphenylhydrazone, carbonyl cyanide 4-trifluoromethoxy phenylhydrazone, 2-2-4-trifluoromethoxy phenyl hydrazono malononitrile, propanedinitrile, 4-trifluoromethoxy phenyl hydrazono, 1-cyano-n-4-trifluoromethoxy phenyl methanecarbohydrazonoyl cyanide, carbonyl cyanide 4-trifluoromethoxyphenylhydrazone, carbonyl cyanide, 4-trifluoromethoxyphenylhydrazone, 4-trifluoromethoxy phenyl hydrazono malononitrile, mesoxalonitrile, p-trifluoromethoxy phenyl hydrazone
C1=CC(=CC=C1NN=C(C#N)C#N)OC(F)(F)F
254.172
CHEBI:75458
>99%
FCCP
C10H5F3N4O
BMZRVOVNUMQTIN-UHFFFAOYSA-N
2-[[4-(trifluoromethoxy)phenyl]hydrazinylidene]propanedinitrile
3330
254.17