missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Fc gamma RIIIA/B (CD16) Antibody, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Fc gamma RIII (CD16) |
---|---|
Clone | 275024 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Fc gamma RIII (CD16) Monoclonal specifically detects Fc gamma RIII (CD16) in Mouse samples. It is validated for Western Blot.Specifications
Fc gamma RIII (CD16) | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q5D5J5 | |
2214 | |
Mouse myeloma cell line NS0-derived recombinant mouse Fc gamma RIII Ala31-Thr215 Accession # Q5D5J5 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Fc gamma RIIIA/B (CD16) in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) Fc gamma RII is observed and no cross-reactivity with rmFc gamma RIA, recombinant human (rh) Fc gamma RIIA, rhFc gamma RIIB/C, rhFc gamma RIIIB, or rmFcrl3 is observed. |
275024 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20 | |
Fcgr3 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only