missing translation for 'onlineSavingsMsg'
Learn More

Fc gamma RIII (CD16) Rat anti-Mouse, Clone: 275024, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1960

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449065

  • £288.60 / 100µg

Estimated Shipment
13-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Fc gamma RIII (CD16) Monoclonal antibody MAB1960 Specifically detects Fc gamma RIII (CD16) in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Fc gamma RIII (CD16)
275024
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Fcgr3
Mouse myeloma cell line NS0-derived recombinant mouse Fc gamma RIII Ala31-Thr215 Accession # Q5D5J5
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2214
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q5D5J5
CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Fc gamma RIIIA/B (CD16) in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) Fc gamma RII is observed and no cross-reactivity with rmFc gamma RIA, recombinant human (rh) Fc gamma RIIA, rhFc gamma RIIB/C, rhFc gamma RIIIB, or rmFcrl3 is observed.
Mouse
Videos
Documents

For Research Use Only