missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Fc gamma RI/CD64 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Fc gamma RI/CD64 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.5 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Fc gamma RI/CD64 Polyclonal specifically detects Fc gamma RI/CD64 in Mouse samples. It is validated for Western Blot.Specifications
Fc gamma RI/CD64 | |
Western Blot 0.5 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
CD64, CD64 antigen, CD64a, CD64b, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FCGR1B, FcgRI, FcgRIA, FCRI, FcRIA, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
Fcgr1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P26151 | |
2209 | |
NS0-derived recombinant mouse Fc gamma RI/CD64 Glu25-Pro297 Accession # P26151 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Fc gamma RI/CD64 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human Fc gamma RI is observed and less than 5% cross-reactivity with recombinant mouse (rm) Fc gamma RIIB and rmFc gamma RIII is observed. |
For Research Use Only