missing translation for 'onlineSavingsMsg'
Learn More

Fc epsilon Receptor 1 alpha (FceRI) Mouse anti-Human, PE-eFluor 610, Clone: AER-37 (CRA1), eBioscience™

Mouse Monoclonal Antibody

£187.00

Specifications

Antigen Fc epsilon Receptor 1 alpha (FceRI)
Clone AER-37 (CRA1)
Host Species Murine
Gene Alias FceRI alpha, high affinity IgE receptor
Applications Flow Cytometry
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15540537
View Documents
Affymetrix eBioscience
61-5899-42
100 Tests
£187.00
100 tests
View alternative products
Save Up to 
»
Estimated Shipment: 02-11-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The AER-37 monoclonal antibody reacts with the Fc epsilon RI alpha subunit, an IgE-binding subunit lacking signal-transducing ability. Fc epsilon RI apha is expressed on mast and basophil cells and is upregulated by the presence of IgE. Fc epsilon RI alpha forms a tetrameric complex with one beta and two gamma subunits. The beta and gamma subunits possess immunoreceptor tyrosine-based activation motifs (ITAM). The Fc epsilon RI complex plays an important role in triggering IgE-mediated allergic reactions.
Specifications

Specifications

Fc epsilon Receptor 1 alpha (FceRI)
Murine
Flow Cytometry
PE-eFluor 610
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Monoclonal
5μL (0.25μg)/test
AER-37 (CRA1)
FceRI alpha, high affinity IgE receptor
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
SDS
Product Certifications

For Research Use Only.