missing translation for 'onlineSavingsMsg'
Learn More

FceR1 alpha, Biotin, clone: AER-37 (CRA1), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5899-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15259919

  • £89.74 / 25µg


Estimated Shipment date
from Supplier 29-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The AER-37 monoclonal antibody reacts with the Fc epsilon RI alpha subunit, an IgE-binding subunit lacking signal-transducing ability. Fc epsilon RI apha is expressed on mast and basophil cells and is upregulated by the presence of IgE. Fc epsilon RI alpha forms a tetrameric complex with one beta and two gamma subunits. The beta and gamma subunits possess immunoreceptor tyrosine-based activation motifs (ITAM). The Fc epsilon RI complex plays an important role in triggering IgE-mediated allergic reactions.Applications Reported: AER-37 has been reported for use in flow cytometric analysis.Applications Tested: The AER-37 (CRA1) antibody has been tested by flow cytometric analysis of peripheral blood cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Fc epsilon Receptor 1 alpha (FceRI)
AER-37 (CRA1)
Biotin
Conjugated
Murine
25μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
FceRI alpha, high affinity IgE receptor
IgG2b
RUO
Primary
Human
Videos