missing translation for 'onlineSavingsMsg'
Learn More

FceR1 alpha, APC, clone: AER-37 (CRA1), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-5899-42

Code : Z2

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 13467846

  • £222.41 / 100 tests


EU Stock 3
Add to basket

Description

Description

Description: The AER-37 monoclonal antibody reacts with the Fc epsilon RI alpha subunit, an IgE-binding subunit lacking signal-transducing ability. Fc epsilon RI apha is expressed on mast and basophil cells and is upregulated by the presence of IgE. Fc epsilon RI alpha forms a tetrameric complex with one beta and two gamma subunits. The beta and gamma subunits possess immunoreceptor tyrosine-based activation motifs (ITAM). The Fc epsilon RI complex plays an important role in triggering IgE-mediated allergic reactions.Applications Reported: The AER-37 (CRA1) antibody has been reported for use in flow cytometric analysis.Applications Tested: This AER-37 (CRA1) antibody has been pre-titrated and tested by flow cytometric analysis of peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Fc epsilon Receptor 1 alpha (FceRI)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P12319
RP11-550P17.3, FCE1A, FcERI
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
AER-37 (CRA1)
APC
FCER1A
Conjugated
FCER1A
IgG2b, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
2205
Videos

For Research Use Only.