Learn More
FceR1 alpha Monoclonal Antibody (MAR-1), PE-Cyanine7, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
£83.00 - £240.00
Specifications
Antigen | FceR1 alpha |
---|---|
Clone | MAR-1 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15518596
|
Affymetrix eBioscience
25-5898-82 |
100 μg |
£240.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15508596
|
Affymetrix eBioscience
25-5898-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The MAR-1 monoclonal antibody reacts with the Fc epsilon Receptor I alpha subunit, an IgE-binding subunit lacking signal-transducing ability. Fc epsilon RI alpha is expressed on mast and basophil cells and is up-regulated by the presence of IgE. Fc epsilon RI alpha forms a tetrameric complex with one beta and two gamma subunits. The beta and gamma subunits possess immunoreceptor tyrosine-based activation motifs (ITAM). The Fc epsilon RI complex plays an important role in triggering IgE-mediated allergic reactions. Applications Reported: This MAR-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MAR-1 antibody has been tested by flow cytometric analysis of MC/9 cell line (a mouse mast cell line). This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
FceR1 alpha Monoclonal antibody specifically detects FceR1 alpha in Mouse samples. It is validated for Flow Cytometry.Specifications
FceR1 alpha | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P20489 | |
IgG | |
Affinity chromatography | |
Antibody |
MAR-1 | |
Flow Cytometry | |
PE-Cyanine7 | |
Armenian Hamster | |
Mouse | |
14125 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.