missing translation for 'onlineSavingsMsg'
Learn More

FceR1 alpha, Biotin, clone: MAR-1, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5898-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15229919

  • £56.04 / 50µg


EU Stock 2
Add to basket

Description

Description

Description: The MAR-1 monoclonal antibody reacts with the Fc epsilon Receptor I alpha subunit, an IgE-binding subunit lacking signal-transducing ability. Fc epsilon RI alpha; is expressed on mast and basophil cells and is up-regulated by the presence of IgE. Fc epsilon RI alpha forms a tetrameric complex with one beta and two gamma subunits. The beta and gamma subunits possess immunoreceptor tyrosine-based activation motifs (ITAM). The Fc epsilon RI complex plays an important role in triggering IgE-mediated allergic reactions.Applications Reported: The MAR-1 antibody has been reported for use in flow cytometric analysis.Applications Tested: The MAR-1 antibody has been tested by flow cytometric analysis of the MC/9 cell line (a mouse mast cell line). This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Fc epsilon Receptor 1 alpha (FceRI)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P20489
RP11-550P17.3, FCE1A, FcERI
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
MAR-1
Biotin
Fcer1a
Conjugated
Fcer1a
IgG
50μg
Store at 2-8°C. Do not freeze.
Primary
14125
Videos

For Research Use Only.