Learn More
FBXL13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563417
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GNKRVTDASF KFIDKNYPNL SHIYMADCKG ITDSSLRSLS PLKQLTVLNL ANCVRIGDMG LKQFLDGPAS MRIRELNLSN CV Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 67%.
Substrate-recognition component of the SCF (SKP1-CUL1-F- box protein)-type E3 ubiquitin ligase complex.Specifications
FBXL13 | |
Polyclonal | |
Unconjugated | |
FBXL13 | |
4921539K22Rik; Drc6; Dynein regulatory complex subunit 6; Fbl13; F-box and leucine rich repeat protein 13; F-box and leucine-rich repeat protein 13; F-box and leucine-rich repeat protein 13-like; F-box/LRR-repeat protein 13; Fbxl13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
222235 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NEE6 | |
FBXL13 | |
Recombinant protein corresponding to Human FBXL13 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only