missing translation for 'onlineSavingsMsg'
Learn More

Fatostatin A, Tocris Bioscience™

Cell permeable inhibitor of SREBP activation

Brand:  RD Systems (Bio-Techne) 4444/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13434774

  • £461.58 / 50mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of sterol regulatory element binding protein (SREBP); impairs the activation of SREBP-1 and SREBP-2. Exhibits antiproliferative effects in DU 145 cells independently of IGF-1 signaling (IC50 = 0.1 μM); reverses hyperglycemia in diabetic (ob/ob) mice. Cell permeable.
Specifications

Chemical Identifiers

298197-04-3
375.328
fatostatin a, 2-2-propylpyridin-4-yl-4-p-tolyl thiazole hydrobromide, 2-propyl-4-4-p-tolyl-thiazol-2-yl-pyridine, fatostatin hydrobromide, fatostatin a hydrobromide, 4-4-4-methylphenyl-2-thiazolyl-2-propylpyridine hydrobromide, 4-4-methylphenyl-2-2-propylpyridin-4-yl-1,3-thiazole hydrobromide, 4-4-4-methylphenyl-1,3-thiazol-2-yl-2-propylpyridine hydrobromide
4-(4-methylphenyl)-2-(2-propylpyridin-4-yl)-1,3-thiazole;hydrobromide
C18H19BrN2S
RJCFNQZVFUMORB-UHFFFAOYSA-N
2850562
CCCC1=NC=CC(=C1)C2=NC(=CS2)C3=CC=C(C=C3)C.Br

Specifications

50mg
298197-04-3
fatostatin a, 2-2-propylpyridin-4-yl-4-p-tolyl thiazole hydrobromide, 2-propyl-4-4-p-tolyl-thiazol-2-yl-pyridine, fatostatin hydrobromide, fatostatin a hydrobromide, 4-4-4-methylphenyl-2-thiazolyl-2-propylpyridine hydrobromide, 4-4-methylphenyl-2-2-propylpyridin-4-yl-1,3-thiazole hydrobromide, 4-4-4-methylphenyl-1,3-thiazol-2-yl-2-propylpyridine hydrobromide
CCCC1=NC=CC(=C1)C2=NC(=CS2)C3=CC=C(C=C3)C.Br
375.328
375.33
Fatostatin A
C18H19BrN2S
RJCFNQZVFUMORB-UHFFFAOYSA-N
4-(4-methylphenyl)-2-(2-propylpyridin-4-yl)-1,3-thiazole;hydrobromide
2850562
>97%
Videos
Documents