missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fasudil dihydrochloride, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
500 mg
1 g
Description
Fasudil dihydrochloride is used in the inhibition of contractile responses to potassium chloride, phenylephnne (PHE), prostaglandin and hepatic stellate cells spreading. It is also used in the formation of stress fibers and in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Fasudil dihydrochloride is used in the inhibition of contractile responses to potassium chloride, phenylephnne (PHE), prostaglandin and hepatic stellate cells spreading. It is also used in the formation of stress fibers and in medicine.
Solubility
Soluble in water and dimethylsulfoxide.
Notes
Incompatible with strong oxidizing agents. Sore in a cool place.
Fasudil dihydrochloride is used in the inhibition of contractile responses to potassium chloride, phenylephnne (PHE), prostaglandin and hepatic stellate cells spreading. It is also used in the formation of stress fibers and in medicine.
Solubility
Soluble in water and dimethylsulfoxide.
Notes
Incompatible with strong oxidizing agents. Sore in a cool place.
Specifications
Color | White |
Quantity | 1 g |
Merck Index | 14,3942 |
Synonym | fasudil dihydrochloride, ha-1077 dihydrochloride, ha 1077, dihydrochloride, 5-1,4-diazepan-1-yl sulfonyl isoquinoline dihydrochloride, 5-isoquinolinesulfonyl homopiperazine, 2hcl, 5-isoquinolinesulfonyl homopiperazine,2hcl, 1-5-isoquinolinylsulfonyl homopiperazine dihydrochloride, 1-5-isoquinoline-sulfonyl-homopiperazine, fasudil and ha 1077, dihydrochloride, ha-1077 dihydrochloride, solid hplc |
Solubility Information | Soluble in water and dimethylsulfoxide. |
Formula Weight | 364.29 |
Percent Purity | ≥99% |
Physical Form | Powder |
Chemical Name or Material | Fasudil dihydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'