missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FASTK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556800
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GQAASSATTR DPAQRVVLVL RERWHFCRDG RVLLGSRALR ERHLGLMGYQ LLPLPFEELE SQRGLPQLKS YLRQKLQAL Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 33%.
FASTK Polyclonal antibody specifically detects FASTK in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
FASTK | |
Polyclonal | |
Unconjugated | |
FASTK | |
0610011K02Rik; 610011K02Rik; Fas activated serine/threonine kinase; Fas-activated serine/threonine kinase; FAST; FAST kinase; Fastk | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10922 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14296 | |
FASTK | |
Recombinant protein corresponding to Human FASTK | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only