missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fas Ligand/TNFSF6 Mouse anti-Rat, Clone: 252029, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Fas Ligand/TNFSF6 |
---|---|
Clone | 252029 |
Applications | Western Blot, Immunocytochemistry |
Conjugate | Unconjugated |
Form | Purified |
Description
Fas Ligand/TNFSF6 Monoclonal specifically detects Fas Ligand/TNFSF6 in Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Fas Ligand/TNFSF6 | |
Western Blot, Immunocytochemistry | |
Purified | |
RUO | |
356 | |
Monoclonal | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
252029 | |
Unconjugated | |
Mouse | |
Rat | |
Mouse myeloma cell line NS0-derived recombinant rat Fas Ligand/TNFSF6 Leu104-Leu278 Accession # P36940 | |
Primary |
For Research Use Only