Learn More
FARS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555988
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: CYFPFTHPSF EMEINFHGEW LEVLGCGVME QQLVNSAGAQ DRIGWAFGLG LERLAMILYD IPDIRLFWCE DERFLKQFCV SNINQKVKFQ PLSKYPAVIN DISFWLPSEN YAEND Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS2 is a phenylalanine-tRNA synthetase (PheRS) localized to the mitochondrion which consists of a single polypeptide chain, unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and in the eukaryotic cytoplasm. Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. FARS1 encodes a mitochondrially-located phenylalanine-tRNA synthetase (PheRS) which consists of a single polypeptide chain unlike the (alpha-beta)2 structure of the prokaryotic and eukaryotic cytoplasmic forms of PheRS. Structure analysis and catalytic properties indicate mitochondrial PheRSs may constitute a class of PheRS distinct from the enzymes found in prokaryotes and the eukaryotic cytoplasm.Specifications
FARS2 | |
Polyclonal | |
Unconjugated | |
FARS2 | |
COXPD14; dJ236A3.1 (phenylalanine-tRNA synthetase); dJ520B18.2; dJ520B18.2 (FARS1 (phenylalanine-tRNA synthetase)); Fars1; FARS2; HSPC320; mitochondrial PHERS; pheny; phenylalanine translase; phenylalanine tRNA ligase 2, mitochondrial; phenylalanine--tRNA ligase, mitochondrial; phenylalanine-tRNA synthetase 1 (mitochondrial); phenylalanine-tRNA synthetase 2 (mitochondrial); Phenylalanyl-tRNA synthetase; phenylalanyl-tRNA synthetase 2, mitochondrial; phenylalanyl-tRNA synthetase, mitochondrial; phenylalanyl-tRNA synthetase, mitochondrial; phenylalanine--tRNA ligase, mitochondrial; pheRS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10667 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95363 | |
FARS2 | |
Recombinant protein corresponding to Human FARS2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only