Learn More
FAP Monoclonal Antibody (F11-24), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience BMS168
151.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: BMS168 (F11-24) specifically reacts with Fibroblast Activation Protein (FAP), a cell surface glycoprotein of reactive human fibroblast. Among the normal adult organs tested BMS168 does not show immunoreactivity in immunohistological studies with either resting fibroblasts (fibrocytes) of the normal organ mesenchyme or the parenchymal cells. The single exception noted is a subset of endocrine cells in the pancreatic islets, presumably glucagon-producing A cells. Applications Tested: Flow Cytometry, Immunohistochemistry, Immunoprecipitation.
FAP Monoclonal antibody specifically detects FAP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation.Specifications
FAP | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q12884 | |
FAP | |
Rec. Human FAP | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
F11-24 | |
Unconjugated | |
FAP | |
170 kDa melanoma membrane-bound gelatinase; Antiplasmin-cleaving enzyme FAP, soluble form; APCE; Dipeptidyl peptidase FAP; DPPIV; Fap; FAPA; FAPalpha; fibroblast activation protein; fibroblast activation protein alpha; fibroblast activation protein, alpha; gelatine degradation protease FAP; Integral membrane serine protease; post-proline cleaving enzyme; Prolyl endopeptidase FAP; seprase; Serine integral membrane protease; SIMP; Surface-expressed protease | |
Mouse | |
Protein G | |
RUO | |
2191 | |
4° C | |
Liquid |
For Research Use Only.