missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM9B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557171
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EEEEGEEEEL IRIFQEQQKK WQQYRSVRRE RLKEMKLLRD QFVKALEDFE DLCDRVFSDE DSEL Highest antigen sequence identity to the following orthologs: Mouse - 44%, Rat - 45%.
FAM9B Polyclonal antibody specifically detects FAM9B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
FAM9B | |
Polyclonal | |
Unconjugated | |
FAM9B | |
FAM9B; family with sequence similarity 9 member B; family with sequence similarity 9, member B; protein FAM9B; testis expressed 39B; TEX39B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
171483 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IZU0 | |
FAM9B | |
Recombinant protein corresponding to Human FAM9B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only