missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse FAM3D Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | FAM3D |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
FAM3D Polyclonal specifically detects FAM3D in Mouse samples. It is validated for Western Blot.Specifications
FAM3D | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
cytokine-like protein EF-7, EF7, family with sequence similarity 3, member D, OIT1 | |
FAM3D | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P97805 | |
131177 | |
Mouse myeloma cell line NS0-derived recombinant mouse FAM3D Trp39-Met224 Accession # P97805 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse FAM3D in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human (rh) FAM3D is observed and less than 1% cross reactivity with rhFAM3A, rhFAM3B and rhFAM3C is observed. |
For Research Use Only