missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM3A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566052
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: GFPRIQQLFTS PESSVTAAPR ARKYKCGLPQ PCPEEHLAFR VVSGAANVIG Highest antigen sequence identity to the following orthologs - mouse 88%, rat 90%.
FAM3A Polyclonal antibody specifically detects FAM3A in Human samples. It is validated for Immunocytochemistry.Specifications
FAM3A | |
Polyclonal | |
Unconjugated | |
FAM3A | |
1810037C20Rik; 2.19; 2-19; Cytokine-like protein 2-19; DLD; DXS560S; FAM3 metabolism regulating signaling molecule A; FAM3A; family 3, member A protein; family with sequence similarity 3 member A; family with sequence similarity 3, member A; hypothetical protein MGC73350; protein FAM3A; RGD1562076; XAP-7; zgc:73350 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
60343 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P98173 | |
FAM3A | |
Recombinant Human FAM3A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only