Learn More
FAM171A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA562386
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LGKDYHKSVE VFPLKARKSM EREGYESSGN DDYRGSYNTV LSQPLFEKQD REGPASTGSK LTIQEHLYPA PSSPEKEQLL DR Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 79%.
FAM171A1 Polyclonal antibody specifically detects FAM171A1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
FAM171A1 | |
Polyclonal | |
Unconjugated | |
FAM171A1 | |
9630050M13Rik; AA409679; APCN; Astroprincin; C10orf38; FAM171A1; family with sequence similarity 171 member A1; family with sequence similarity 171, member A1; protein FAM171A1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
221061 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VUB5 | |
FAM171A1 | |
Recombinant protein corresponding to Human FAM171A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only