missing translation for 'onlineSavingsMsg'
Learn More

FAM160A1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA561221

Additional Details : Weight : 0.02000kg

Product Code. 15739041

  • £441.00 / 100µL
Estimated Shipment: 09-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: LLHHKPILKP LMMLLSSCSG TTTPTVEEKL VVLLNQLCSI LAKDPSILEL FFHTSEDQGA ANFLIFS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

FAM160A1 Polyclonal antibody specifically detects FAM160A1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

FAM160A1
Polyclonal
Unconjugated
FAM160A1
FAM160A1; family with sequence similarity 160 member A1; family with sequence similarity 160, member A1; protein FAM160A1
Rabbit
Antigen affinity chromatography
RUO
729830
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q05DH4
FAM160A1
Recombinant protein corresponding to Human FAM160A1
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only