missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM149B1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA558953
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DDKLLMRPGS STILSTRNWP NRAVEFSTSS LSYTVQSTRR RNPPPRTLHP ISTSHSCAET PRSVEEILRG ARVPVAPDSL SSPSP Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 79%.
FAM149B1 Polyclonal antibody specifically detects FAM149B1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
FAM149B1 | |
Polyclonal | |
Unconjugated | |
FAM149B1 | |
FAM149B1; family with sequence similarity 149 member B1; family with sequence similarity 149, member B1; KIAA0974; Protein FAM149B1; RGD1359592 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
317662 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96BN6 | |
FAM149B1 | |
Recombinant protein corresponding to Human FAM149B1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only