missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM13B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567216
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: YSLLKFLCRFL ANVASHHEEI WSANSLAAVF GPDVFHIYTD VEDMKEQEIV SRIMAGLLEN YYEFFENEEE DFSSNDLSSI Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
FAM13B Polyclonal antibody specifically detects FAM13B in Human samples. It is validated for Immunohistochemistry.Specifications
FAM13B | |
Polyclonal | |
Unconjugated | |
FAM13B | |
2610024E20Rik; ARHGAP49; AW060714; AW546153; C5orf5; Fam13b; FAM13B1; family with sequence similarity 13 member B; family with sequence similarity 13, member B; family with sequence similarity 13, member B1; GAP-like protein N61; KHCHP; N61; protein FAM13B; RGD1310484 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51306 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NYF5 | |
FAM13B | |
Recombinant Human FAM13B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only