Learn More
FAM134A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA552801
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SGGFSVSRAT TPQLTDVSED LDQQSLPSEP EETLSRDLGE GEEGELAPPE DLLGRPQALS RQALDSEEEE EDVAAKETLL RLSSPLHFVN THFNGAGSPP DGVKCSPGGP VETLSPETVS GGLTALPGTL SPPLCLVGSD PA Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 80%.
FAM134A Polyclonal antibody specifically detects FAM134A in Human samples. It is validated for Immunohistochemistry.Specifications
FAM134A | |
Polyclonal | |
Unconjugated | |
RETREG2 | |
C2H2orf17; C2orf17; Fam134a; family with sequence similarity 134 member A; family with sequence similarity 134, member A; MAG2; MAG-2; metastasis associated gene-2; protein FAM134A; reticulophagy regulator 2; reticulophagy regulator 2; protein FAM134A; reticulophagy regulator family member 2; Retreg2; RGD1306844; zgc:158472 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79137 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NC44 | |
RETREG2 | |
Recombinant protein corresponding to Human FAM134A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only