Learn More
FAM122B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA551371
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EPDTSYGGTL RRSSSAPLIH GLSDLSQVFQ PYTLRTRRNS TTIMSRHSLE EGLDMVNRET AHEREMQTAM QISQSWDESL SLSDSDFDKP EKLYSPKRID FTPVSPAPSP TRGFGKMFVS SS Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.
FAM122B Polyclonal antibody specifically detects FAM122B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
FAM122B | |
Polyclonal | |
Unconjugated | |
FAM122B | |
4632404H22Rik; AI256344; erg-3; ets-related; FAM122B; family with sequence similarity 122, member B; family with sequence similarity 122B; p55; protein FAM122B; RP11-308B5.5; SPACIA2; synoviocyte proliferation associated in collagen-induced arthritis 2; TMPRSS2/ERG fusion | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
159090 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z309 | |
FAM122B | |
Recombinant protein corresponding to Human FAM122B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only