missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM118B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA559867
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KHKSDLEHFM LVRRGDVDEF KKLRENMLDK GIKVISYGDD YADLPEYFKR LTCEISTRGT SAGMVREGQL NGSSAAHSE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
FAM118B Polyclonal antibody specifically detects FAM118B in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
FAM118B | |
Polyclonal | |
Unconjugated | |
FAM118B | |
2310022O21Rik; 2700018L24Rik; C030004A17Rik; Fam118b; family with sequence similarity 118 member B; family with sequence similarity 118, member B; Ppp4r1l; protein FAM118B; RGD1307173 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79607 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BPY3 | |
FAM118B | |
Recombinant protein corresponding to Human FAM118B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only