Learn More
FAM107B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA558871
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FHASIPRPSI IDTPKEEEFR EEPKCLELEQ KMTSDSPPED IDHKDSYLIT RSIMAEPDYI EDDNPELIRP QKLINPVK Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 49%.
FAM107B Polyclonal antibody specifically detects FAM107B in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
FAM107B | |
Polyclonal | |
Unconjugated | |
FAM107B | |
3110001A13Rik; AA589595; AA959793; AI132312; axoneme-associated protein mst101(2)-like; C10orf45; Fam107b; fam107b protein; family with sequence similarity 107 member B; family with sequence similarity 107, member B; FLJ45505; HITS; hypothetical protein LOC386641; hypothetical protein MGC11034-like protein; MGC11034; MGC137524 protein; novel DUF1151 domain containing protein; protein FAM107B; wu:fc52f02; zgc:56216 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83641 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H098 | |
FAM107B | |
Recombinant protein corresponding to Human FAM107B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only