missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ Polystyrene Microplates

Constructed of crystal grade virgin polystyrene for optimum visibility. Corning™ Falcon™ Polystyrene Microplates are available with tissue culture-treated and nontreated surfaces.

Brand:  Falcon 353077

Code : 66

Additional Details : Weight : 3.80000kg

 View more versions of this product

Product Code. 10777241

  • £134.00 / Pack of 50


In Stock 12
Add to basket

Description

Description

  • Choose tissue culture-treated or nontreated surfaces
  • Nontreated growth surface is more hydrophobic than tissue culture surface and is similiar to the surface of a bacteriological Petri dish
  • Sterilized by gamma irradiation
  • Lid design permits stable stacking on benchtop or in incubator
  • All plates (except 08-772-2A and 08-772-16) have patented “labyrinth” lid that reduces evaporation of media from wells, minimizes the risk of contamination
  • Deep skirt and serrated edge permit a firmer grip for plate manipulation
  • Exceptional optical clarity allows for visualization of all cell monolayers
  • Frosted writing ends provide clear writing area for quick identification
  • All plates and lids are suitable for both manual and automated handling

Specifications

Specifications

Cell Culture Microplate
Clear
96 well
96
Sterile
1.5 to 2.2mL
Round
U shaped
0.36cm2
Polystyrene
1/Tray, 50/Cs.
Tissue-culture treated
With Lid
0.32mL
Videos