Learn More
Falcon™ Plates in Ready-Stack Packaging
Trays are recyclable, stackable, and reusable for transporting, storing, or incubating plates
£143.00 - £248.62
Specifications
Sterility Sterile
Material Polystyrene
View More Specs
Includes: Lids
Products 2
Product Code Brand Culture Area Format Volume (Metric) No. of Wells Price Quantity & Availability  
Product Code Brand Culture Area Format Volume (Metric) No. of Wells Price Quantity & Availability  
10021202 View Documents Falcon
353935
2 cm2 3.5 mL 24 £143.00
Pack of 60 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
10413972 View Documents Falcon
353936
0.32 cm2 0.37mL 96 £248.62
Pack of 84 View alternative products
Save Up to 
»
Estimated Shipment
18-10-2019
Description
Description
  • Bulk packaged in medical-style peel-open trays for convenient, aseptic handling
  • Plates positioned on their sides, permitting labeling before removal
  • Tissue culture-treated
Order Info
Packaged sterile, 14 plates per tray
Specifications
Specifications
Sterile
Polystyrene