Learn More
FAK Inhibitor 14, Tocris Bioscience™
Selective FAK inhibitor
Brand:  Tocris (Bio-Techne) 341450
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13462974
  • £480.69 / 50mg
Estimated Shipment date
from Supplier 24-03-2020
Description
Description
Selective focal adhesion kinase (FAK) inhibitor that displays no significant activity at a range of other kinases including EGFR, PDGFR and IGF-RI. Prevents FAK autophosphorylation at Y397 (IC50 = 1 μM), promotes cell detachment and inhibits cell adhesion in vitro. Exhibits antiproliferative activity in a variety of human tumor cell lines in vitro and in breast cancer cells in vivo.
Specifications
Chemical Identifiers
4506-66-5
284.006
benzene-1,2,4,5-tetraamine tetrahydrochloride, 1,2,4,5-benzenetetramine tetrahydrochloride, fak inhibitor 14, 1,2,4,5-benzenetetramine, tetrahydrochloride, 1,2,4,5-tetraaminobenzene tetrahydrochloride, benzene-1,2,4,5-tetrayltetraamine tetrahydrochloride, benzene-1,2,4,5-tetramine 4hcl, benzene-1,2,4,5-tetramine tetrahydrochloride, 1,2,4,5-benzenetetraamine tetrahydrochloride, tabh
benzene-1,2,4,5-tetramine;tetrahydrochloride
C6H14Cl4N4
BZDGCIJWPWHAOF-UHFFFAOYSA-N
78260
C1=C(C(=CC(=C1N)N)N)N.Cl.Cl.Cl.Cl
Specifications
50mg
4506-66-5
benzene-1,2,4,5-tetraamine tetrahydrochloride, 1,2,4,5-benzenetetramine tetrahydrochloride, fak inhibitor 14, 1,2,4,5-benzenetetramine, tetrahydrochloride, 1,2,4,5-tetraaminobenzene tetrahydrochloride, benzene-1,2,4,5-tetrayltetraamine tetrahydrochloride, benzene-1,2,4,5-tetramine 4hcl, benzene-1,2,4,5-tetramine tetrahydrochloride, 1,2,4,5-benzenetetraamine tetrahydrochloride, tabh
C1=C(C(=CC(=C1N)N)N)N.Cl.Cl.Cl.Cl
284.006
284.01
FAK Inhibitor 14
C6H14Cl4N4
BZDGCIJWPWHAOF-UHFFFAOYSA-N
benzene-1,2,4,5-tetramine;tetrahydrochloride
78260
>98%