missing translation for 'onlineSavingsMsg'
Learn More

Logistik Unicorp™ Faithful White Color 99% Polyester 1% Carbon Lab Coat

Catalog Number 12872895
Click to view available options
Size (Metric) Chest:
92cm
100cm
108cm
This item is not returnable. View return policy
This item is not returnable. View return policy
Type Lab Coat
Size (Metric) Chest 92cm
Includes Two side pockets, Nickel-free studded front c/w extended flap over, stud cuffs
Weight (Metric) 270 g/m2
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'