missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Logistik Unicorp™ Faithful White Color 65% Polyester/35% Cotton Lab Coat
Protect those working in the lab with a Logistik Unicorp™ Faithful White Color 65% Polyester/35% Cotton Lab Coat. The white coat comes in four unisex sizes, is not CE marked and conforms to Howie report guidelines.
£41.85 - £79.75
Product Code | Brand | Size (Metric) Chest | No. of Pockets | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size (Metric) Chest | No. of Pockets | Price | Quantity & Availability | ||||
12951275
|
Logistik Unicorp™
441AAA-V6WH-108 |
108cm | 2 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12844382
|
Logistik Unicorp™
849A-E2WH-116CM |
116cm | 3 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12961275
|
Logistik Unicorp™
441AAA-V6WH-116 |
116cm | 2 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12811141
|
Logistik Unicorp™
441-V6WH-132 |
132cm | 3 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Conforms to Howie report guidelines
- Side fastening
- Stainless steel stud fastening at wrist
- Four unisex sizes
- Polyester cotton;
- Not CE marked
Specifications
Lab Coat | |
White |